N-Benzyl-3-borono-4-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBBB-2612-1GEA 796 EUR
COBBBB-2612-1G
N-Benzyl-3-borono-4-methylbenzolsulfonamid
N-Benzyl-3-borono-4-methylbenzolsulfonamid
Formel: C₁₄H₁₆BNO₄S
Molecular Weight: 305,16 g/mol
MDL: MFCD07363760
CAS-Nummer: 871329-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR