2-Amino-5-methylbenzolsulfonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL218766-250GEA 59.4 EUR
SIAL218766-250G
2-Amino-5-methylbenzolsulfonsäure, Sigma-Aldrich®
2-Amino-5-methylbenzenesulphonic acid
Formel: C₇H₉NO₃S
Molecular Weight: 187,22
MDL: MFCD00007908
CAS-Nummer: 88-44-8
EINECS: 201-831-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR