2-Amino-5-methylbenzolsulfonsäure
Lieferant: EHRENSTORFER GMBH
EHERC10228000EA
39
EUR
EHERC10228000
2-Amino-5-methylbenzolsulfonsäure
2-Amino-5-methylbenzenesulphonic acid
Formel:
C₇H₉NO₃S Molecular Weight: 187,22 g/mol |
MDL:
MFCD00007908 CAS-Nummer: 88-44-8 EINECS: 201-831-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...