4'-(Dimethylamino)-2,2,2-trifluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL514667-1GEA 122 EUR
SIAL514667-1G
4'-(Dimethylamino)-2,2,2-trifluoracetophenon, Sigma-Aldrich®
4'-(Dimethylamino)-2,2,2-trifluoracetophenon
Formel: C₁₀H₁₀F₃NO
MDL: MFCD00099468
CAS-Nummer: 2396-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR