2,2,2-Trifluoracetophenon-4-boronsäurepinakolester

Lieferant: FRONTIER SCIENTIFIC INC

FRONT12462.1GEA 0 EUR
FRONT12462.1G
2,2,2-Trifluoracetophenon-4-boronsäurepinakolester
2,2,2-Trifluoracetophenon-4-boronsäurepinakolester
Formel: C₁₄H₁₆BF₃O₃
Molecular Weight: 300,0813 g/mol
MDL: MFCD16628195
CAS-Nummer: 1004294-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR