2-Brom-5'-fluor-2'-hydroxyacetophenone, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL663581-5GEA 242 EUR
SIAL663581-5G SIAL663581-1G
2-Brom-5'-fluor-2'-hydroxyacetophenone, Sigma-Aldrich®
2-Brom-5'-fluor-2'-hydroxyacetophenone
Formel: C₈H₆BrFO₂
MDL: MFCD03094564
CAS-Nummer: 126581-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR