2-Brom-2',6'-dichlor-3'-fluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL669415-1GEA 30.3 EUR
SIAL669415-1G
2-Brom-2',6'-dichlor-3'-fluoracetophenon, Sigma-Aldrich®
2-Brom-2',6'-dichlor-3'-fluoracetophenon
Formel: C₈H₄BrCl₂FO
MDL: MFCD09038425
CAS-Nummer: 914635-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR