2-Methoxy-5-(trifluormethoxy)anilin ≥97%

Lieferant: Thermo Scientific
Gefahr

H32219.03EA 85.3 EUR
H32219.03 H32219.06
2-Methoxy-5-(trifluormethoxy)anilin ≥97%
2-Methoxy-5-(trifluormethoxy)anilin
Formel: C₈H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832305
CAS-Nummer: 660848-57-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR