2-Fluor-3-formylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8091-1GEA 47.4 EUR
COBBBB-8091-1G
2-Fluor-3-formylphenylboronsäure
2-Fluor-3-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,9300 g/mol
MDL: MFCD07369725
CAS-Nummer: 849061-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR