2,6-Difluor-4-formylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8736-1GEA 135 EUR
COBBPN-8736-1G COBBPN-8736-250M
2,6-Difluor-4-formylphenylboronsäurepinakolester
2,6-Difluor-4-formylphenylboronsäurepinakolester
Formel: C₁₃H₁₅BF₂O₃
MDL: MFCD08276848
CAS-Nummer: 870717-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR