2-Chlor-4-fluor-5-nitrophenol

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT11L-319S.10EA 0 EUR
BIOT11L-319S.10 CHMP11L-319S.005 BIOT11L-319S.5 BIOT11L-319S.500 BIOT11L-319S.1
2-Chlor-4-fluor-5-nitrophenol
2-Chlor-4-fluor-5-nitrophenol
Formel: C₆H₃ClFNO₃
MDL: MFCD02670258
CAS-Nummer: 84478-75-1
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR