5-Chlor-2-formylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-9014-1GEA 160 EUR
COBBBB-9014-1G
5-Chlor-2-formylphenylboronsäure
5-Chlor-2-formylphenylboronsäure
Formel: C₇H₆BClO₃
MDL: MFCD03425955
CAS-Nummer: 870238-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR