3-Chlor-4-formylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8545-1GEA 270 EUR
COBBBB-8545-1G
3-Chlor-4-formylphenylboronsäure
3-Chlor-4-formylphenylboronsäure
Formel: C₇H₆BClO₃
Molecular Weight: 184,39 g/mol
MDL: MFCD08274473
CAS-Nummer: 1072952-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR