N-Butyl-4-boronobenzolsulfonamid ≥97%
|
Achtung
|
H53218.03EA
111
EUR
H53218.03
N-Butyl-4-boronobenzolsulfonamid ≥97%
N-Butyl-4-boronobenzolsulfonamid
Formel:
C₁₀H₁₆BNO₄S Molecular Weight: 257,11 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363753 CAS-Nummer: 850589-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...