N-Isopropyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBBB-2616-1GEA 478 EUR
COBBBB-2616-1G
N-Isopropyl-3-boronobenzolsulfonamid
N-Isopropyl-3-boronobenzolsulfonamid
Formel: C₉H₁₄BNO₄S
Molecular Weight: 243,08 g/mol
MDL: MFCD07783856
CAS-Nummer: 871329-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR