4-(N,N-Diphenylamino)phenylboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBBB-8149-1GEA
28
EUR
COBBBB-8149-1G
COBBBB-8149-5G
4-(N,N-Diphenylamino)phenylboronsäure
4-(N,N-Diphenylamino)phenylboronsäure
Formel:
C₁₈H₁₆BNO₂ Molecular Weight: 289,14 |
MDL:
MFCD06798117 CAS-Nummer: 201802-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...