3-(1,3-Oxazol-5-yl)anilin

Lieferant: CHEMPUR

CHMPSEW00990.10EA 0 EUR
CHMPSEW00990.10 MAYBSEW00990DA
3-(1,3-Oxazol-5-yl)anilin
3-(1,3-Oxazol-5-yl)anilin
Formel: C₉H₈N₂O
MDL: MFCD00052191
CAS-Nummer: 157837-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR