3'-Allyl-4'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL632759-1GEA 10.8 EUR
SIAL632759-1G SIAL632759-5G
3'-Allyl-4'-hydroxyacetophenon, Sigma-Aldrich®
3'-Allyl-4'-hydroxyacetophenon
Formel: C₁₁H₁₂O₂
MDL: MFCD00068015
CAS-Nummer: 1132-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR