Tris[N,N-bis(trimethylsilyl)amid]cer(III) ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
44362.14EA
1797
EUR
44362.14
ABCRAB204447.0001
Tris[N,N-bis(trimethylsilyl)amid]cer(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amid]cer(III)
Formel:
C₁₈H₅₄CeN₃Si₆ Molecular Weight: 621,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411241 CAS-Nummer: 41836-21-9 UN: 3396 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...