2,4-Dimethoxy-5-pyrimidinboronsäure

Lieferant: CHEMPUR

CHMPFL11033.5EA 0 EUR
CHMPFL11033.5 FLUO011033.5G FLUO011033.1G
2,4-Dimethoxy-5-pyrimidinboronsäure
2,4-Dimethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₄
Molecular Weight: 183,9577 g/mol
MDL: MFCD01318985
CAS-Nummer: 89641-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR