5-Pyrimidinboronsäure
Lieferant: Thermo Scientific
MAYBCC07412DAEA 0 EUR
MAYBCC07412DA
5-Pyrimidinboronsäure
5-Pyrimidineboronic acid
Formel: C₄H₅BN₂O₂
Molecular Weight: 123,91 g/mol
MDL: MFCD03002366
CAS-Nummer: 109299-78-7