5-Pyrimidinboronsäure

Lieferant: ABCR GMBH

ABCRAB175517.0002EA 0 EUR
ABCRAB175517.0002
5-Pyrimidinboronsäure
5-Pyrimidineboronic acid
Formel: C₄H₅BN₂O₂
Molecular Weight: 123,91
MDL: MFCD03002366
CAS-Nummer: 109299-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR