2-(Cyclopentylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-5933-1GEA 347 EUR
COBBPN-5933-1G
2-(Cyclopentylamino)-5-pyrimidinboronsäurepinakolester
2-(Cyclopentylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₅H₂₄BN₃O₂
MDL: MFCD11878346
CAS-Nummer: 1202805-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR