2-(Diethylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-5922-1GEA 571 EUR
COBBPN-5922-1G
2-(Diethylamino)-5-pyrimidinboronsäurepinakolester
2-(Diethylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₄BN₃O₂
Molecular Weight: 277,1700 g/mol
MDL: MFCD12546555
CAS-Nummer: 1218791-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR