4-Brom-1,3,5-trimethyl-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL647667-1GEA 41.6 EUR
SIAL647667-1G SIAL647667-5G
4-Brom-1,3,5-trimethyl-1H-pyrazol, Sigma-Aldrich®
4-Bromo-1,3,5-trimethyl-1H-pyrazole
Formel: C₆H₉BrN₂
Molecular Weight: 189,06
MDL: MFCD00052532
CAS-Nummer: 15801-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR