5-tert-Butyl-3-iod-1H-pyrazol
Lieferant: COMBI-BLOCKS INC.
COBBHI-1646-250MEA
275
EUR
COBBHI-1646-250M
COBBHI-1646-1G
5-tert-Butyl-3-iod-1H-pyrazol
5-tert-Butyl-3-iod-1H-pyrazol
Formel:
C₇H₁₁IN₂ |
MDL:
MFCD13195722 CAS-Nummer: 1218791-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...