3-Amino-4-brom-1-methyl-1H-pyrazol

Lieferant: CHEMPUR

CHMPGK00323.1EA 0 EUR
CHMPGK00323.1 CHMPGK00323.250
3-Amino-4-brom-1-methyl-1H-pyrazol
3-Amino-4-bromo-1-methyl-1H-pyrazole
Formel: C₄H₆BrN₃
Molecular Weight: 176,02 g/mol
MDL: MFCD00663065
CAS-Nummer: 146941-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR