4-Brom-3-methyl-1H-pyrazol
Lieferant: Thermo Scientific
MAYBJFD00652.10EA
0
EUR
MAYBJFD00652.10
4-Brom-3-methyl-1H-pyrazol
4-Bromo-3-methyl-1H-pyrazole
Formel:
C₄H₅BrN₂ Molecular Weight: 161,01 g/mol |
MDL:
MFCD00005241 CAS-Nummer: 13808-64-5 EINECS: 237-456-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...