4-Iod-1-methylimidazol-5-carbaldehyd

Lieferant: COMBI-BLOCKS INC.

COBBHI-1028-1GEA 1430 EUR
COBBHI-1028-1G
4-Iod-1-methylimidazol-5-carbaldehyd
4-Iod-1-methylimidazol-5-carbaldehyd
Formel: C₅H₅IN₂O
MDL: MFCD01632219
CAS-Nummer: 189014-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR