7-Bromacetyl-3,4-dihydro-2H-1,5-benzodioxepin

Lieferant: Thermo Scientific

MAYBBTB12325.0001EA 0 EUR
MAYBBTB12325.0001
7-Bromacetyl-3,4-dihydro-2H-1,5-benzodioxepin
7-Bromacetyl-3,4-dihydro-2H-1,5-benzodioxepin
Formel: C₁₁H₁₁BrO₃
Molecular Weight: 271,11
MDL: MFCD00218462
CAS-Nummer: 35970-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR