1,4-Benzodioxan-6-boronsäure

Lieferant: CHEMPUR

CHMPCC01312.250EA 0 EUR
CHMPCC01312.250
1,4-Benzodioxan-6-boronsäure
1,4-Benzodioxan-6-boronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,97 g/mol
MDL: MFCD01009696
CAS-Nummer: 164014-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR