p-Tolylchlorformat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL328758-5GEA 65.1 EUR
SIAL328758-5G
p-Tolylchlorformat, Sigma-Aldrich®
p-Tolylchlorformat
Formel: C₈H₇ClO₂
MDL: MFCD00013255
CAS-Nummer: 937-62-2
EINECS: 213-332-8
UN: 3277
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR