9,12-Octadecadiinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALO5636-1MGEA 233 EUR
SIALO5636-1MG
9,12-Octadecadiinsäure, Sigma-Aldrich®
9,12-Octadecadiynoic acid
Formel: C₁₈H₂₈O₂
MDL: MFCD00797685
CAS-Nummer: 2012-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR