Allantoin

Lieferant: Molekula

MOLEM56119443EA 0 EUR
MOLEM56119443
Allantoin
Allantoin
Formel: C₄H₆N₄O₃
Molecular Weight: 158,12 g/mol
MDL: MFCD00005260
CAS-Nummer: 97-59-6
EINECS: 202-592-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR