4-(Iodacetyl)benzophenon

Lieferant: Uptima

UPTIBI1351EA 0 EUR
UPTIBI1351
4-(Iodacetyl)benzophenon
4-(Iodacetyl)benzophenon
Formel: C₁₅H₁₁IO₂
CAS-Nummer: 76809-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR