3-(1-Acetoxyethyl)benzophenon

Lieferant: LGC Standards PROMOCHEM

PROOMM0001.50EA 574 EUR
PROOMM0001.50
3-(1-Acetoxyethyl)benzophenon
3-(1-Acetoxyethyl)benzophenon
Formel: C₁₇H₁₆O₃
Molecular Weight: 268,31 g/mol
CAS-Nummer: 2377141-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR