4-Amino-1-Boc-piperidin-4-carbonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL680095-500MGEA
115
EUR
SIAL680095-500MG
4-Amino-1-Boc-piperidin-4-carbonsäure, Sigma-Aldrich®
4-Amino-1-Boc-piperidin-4-carbonsäure
Formel:
C₁₁H₂₀N₂O₄ Molecular Weight: 244,29 |
MDL:
MFCD01318728 CAS-Nummer: 183673-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...