4-Iod-1-(pyridin-4-ylmethyl)-1H-pyrazol
Lieferant: COMBI-BLOCKS INC.
COBBHI-1565-1GEA
196
EUR
COBBHI-1565-1G
COBBHI-1565-5G
4-Iod-1-(pyridin-4-ylmethyl)-1H-pyrazol
4-Iod-1-(pyridin-4-ylmethyl)-1H-pyrazol
Formel:
C₉H₈IN₃ |
MDL:
MFCD12913970 CAS-Nummer: 1187385-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...