1,1,2-Trimethyl-1H-benz[e]indol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL03024-25G-FEA 40 EUR
SIAL03024-25G-F
1,1,2-Trimethyl-1H-benz[e]indol, Sigma-Aldrich®
1,1,2-Trimethyl-1H-benz[e]indol
Formel: C₁₅H₁₅N
Molecular Weight: 209,29
MDL: MFCD00082627
CAS-Nummer: 41532-84-7
EINECS: 255-429-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR