7-(2-Hydroxyethyl)theophyllin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALH9006-25GEA 88.1 EUR
SIALH9006-25G
7-(2-Hydroxyethyl)theophyllin, Sigma-Aldrich®
7-(2-Hydroxyethyl)theophyllin
Formel: C₉H₁₂N₄O₃
MDL: MFCD00055055
CAS-Nummer: 519-37-9
EINECS: 208-269-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR