Theophyllin

Lieferant: U.S. Pharmacopeia

USPH1653117EA 239 EUR
USPH1653117
Theophyllin
Theophyllin
Formel: C₇H₈N₄O₂
MDL: MFCD00079619
CAS-Nummer: 58-55-9
EINECS: 200-385-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR