8-Chlortheophyllin
Lieferant: LGC Standards PROMOCHEM
PROOMM0155.02EA
416
EUR
PROOMM0155.02
8-Chlortheophyllin
8-Chlortheophyllin
Formel:
C₇H₇ClN₄O₂ |
MDL:
MFCD00005581 CAS-Nummer: 85-18-7 EINECS: 201-590-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...