8-Chlortheophyllin

Lieferant: Molekula

MOLE33137250-100GEA 199 EUR
MOLE33137250-100G
8-Chlortheophyllin
8-Chlortheophyllin
Formel: C₇H₇ClN₄O₂
MDL: MFCD00005581
CAS-Nummer: 85-18-7
EINECS: 201-590-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR