(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
Lieferant: Thermo Scientific
MAYBJFD03645.1EA
0
EUR
MAYBJFD03645.1
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
Formel:
C₂₂H₂₂BrO₂P Molecular Weight: 429,30 g/mol |
MDL:
MFCD00011966 CAS-Nummer: 52509-14-5 EINECS: 257-977-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...