Bis(pinakolato)dibor

Lieferant: ABCR GMBH

ABCRAB172302.0025EA 0 EUR
ABCRAB172302.0025
Bis(pinakolato)dibor
Bis(pinakolato)dibor
Formel: C₁₂H₂₄B₂O₄
Molecular Weight: 253,94 g/mol
MDL: MFCD00799570
CAS-Nummer: 73183-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR