Tricyclazol

Lieferant: EHRENSTORFER GMBH

EHERC17810000EA 114 EUR
EHERC17810000
Tricyclazol
Tricyclazol
Formel: C₉H₇N₃S
CAS-Nummer: 41814-78-2
EINECS: 255-559-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR