N(α)-Cbz-L-histidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL408549-5GEA 92.5 EUR
SIAL408549-5G
N(α)-Cbz-L-histidin, Sigma-Aldrich®
N(α)-Cbz-L-histidin
Formel: C₁₄H₁₅N₃O₄
Molecular Weight: 289,3 g/mol
MDL: MFCD00065960
CAS-Nummer: 14997-58-1
EINECS: 239-084-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR