2-(2-Thienyliden)-4'-phenoxyacetophenon

Lieferant: MENAI (PART OF CHEMPUR)

MENACB791.50EA 0 EUR
MENACB791.50
2-(2-Thienyliden)-4'-phenoxyacetophenon
2-(2-Thienyliden)-4'-phenoxyacetophenon
Formel: C₁₉H₁₄O₂S
MDL: MFCD00170623
CAS-Nummer: 333772-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR