Diosmin
Lieferant: U.S. Pharmacopeia
USPH1213318EA
423
EUR
USPH1213318
Diosmin
Diosmin
Formel:
C₂₈H₃₂O₁₅ |
MDL:
MFCD00009772 CAS-Nummer: 520-27-4 EINECS: 208-289-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...