Buffer, bicarbonate, 1M solution, pH 8,0

Lieferant: Thermo Fisher Scientific
J63491.APEA 69.16 EUR
J63491.AP J63491.AK
Buffer, bicarbonate, 1M solution, pH 8,0
Puffer Biologische Puffer
CAS No.: 144-55-8
Physical form: liquid
MDL No.: MFCD00003528
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR